Learn More
Ansell™ HyFlex™ Light Duty Cut-Resistant Nylon Gloves with Thumbslot
Catalog No. 19010257 Shop All Ansell Products
This item is not returnable. View return policy
Catalog No. 19-010-257 Supplier Ansell™ Supplier No. 11271180
Add to Cart
Add to Cart
This item is not returnable. View return policy
Ansell™ HyFlex™ Light Duty Cut-Resistant Nylon Gloves sleeves are double-layered, made of nylon for soft and comfortable wear experience.
  • Seamless design
  • Guage: 24
  • For light mechanical protection
  • Provides an enhanced protection of wrist
  • Wrist and elastic band at top provide stability for forearms
  • Ambidextrous and can be worn by left arm or by right arm
  • Dirt masking colour for prolonged wear
  • Silicone free for use in paint and finish process
    TRUSTED_SUSTAINABILITY
    Adhere to
    Safety Regulations and Best Practices
    Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.