Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell™ HyFlex™ Sleeve with Intercept™ Technology
GREENER_CHOICE

Catalog No. 19010259 Shop All Ansell Products
This item is not returnable. View return policy
Catalog No. 19-010-259 Supplier Ansell™ Supplier No. 11250120N
Add to Cart
Add to Cart
This item is not returnable. View return policy

Ansell™ HyFlex™ Sleeve with Intercept™ Technology features a knitted bicep cuff to keep the sleeve in place

  • Available in different lengths and widths for optimal fit and comfort to fit.
  • Color-coding helps for easy sorting after laundry
  • Made of black yarn
  • Silicone-free for friendly paint and finish process
  • Compliant with FDA food handling regulations 21 CFR 177.1520 and 21 CFR 177.2600
  • Applications:
    • Press shop operations
    • Handling metal sheets and panels
    • Handling parts with sharp, rough edges
    • Handling structural and body components
    • Inspection, selecting, checking parts
    • Glass repair and replacement
    • Seizing structural and body parts
    • Body surface checking, marking and touching up
    • Adjusting systems and screwing
    • Adjusting panels parts components
TRUSTED_SUSTAINABILITY

Specifications

Product Type Cut-resistant Sleeve
Size Narrow
Construction Knitted
Material HPPE Yarn
Sterility Non-sterile
Style Ambidextrous
Disposable Reusable
Closure Type Elastic Upper with Knit Cuff
Seam Type Seamless
Cuff Style Sleeve
Model HyFlex™ 11-250
Certifications/Compliance FDA CFR 177.1520 and 21 CFR 177.2600; EN420:2003 + A1:2009; Category ll
Length (English) 12 in.
Length (Metric) 30 cm
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.