Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hygiena Q-Swab™
SDP

Catalog No. 14037785
Change view
Click to view available options
Tip Material:
Buffered Peptone Water
Letheen Broth
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Tip Material
14-037-785 Buffered Peptone Water
14-037-764 Letheen Broth
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-037-785 Supplier Hygiena Supplier No. QS1000
Only null left
Add to Cart
Add to Cart

All-in-one dilution and delivery device system for environmental sample collection

Once the sample area has been swabbed, snapping the Snap-Valve releases 1mL of broth into swab shaft, neutralizing residual sanitizers and facilitating the recovery of bacteria. The sample is then ready to be poured onto dehydrated media or streaked onto agar plates for culturing.

  • Each broth is gamma irradiated and aseptically filled into a self-contained reservoir at the tip of the device
  • Ideal for surface sampling in the food and beverage industry
  • Eliminate the need for pipettes and broth preparation
  • Swabs can be used either wet or dry
  • 1mL sample perfectly hydrates dried media
  • Neutralizes residual sanitizers remaining on surface
  • Environmental microbial sample collection and transport
  • All-in-one sample collection system
  • 250 devices per case

Specifications

For Use With (Application) Ideal for surface sampling in the food and beverage industry
Storage Requirements 2°C to 8°C
Tip Material Buffered Peptone Water
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.