Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hygromycin B, 50 mg/ml in distilled water, sterile-filtered

Catalog No. AAJ673718EQ
Change view
Click to view available options
Quantity:
1 mL
5 x 1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ673718EQ 5 x 1 mL
AAJ67371XF 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ673718EQ Supplier Thermo Scientific Chemicals Supplier No. J673718EQ
Add to Cart

CAS: 31282-04-9 | C20H37N3O13 | 527.524 g/mol

Hygromycin B was used for hygro resistance effect in the following cells transfected 293T cells with mixture of plasmids at 160 ?g/ml, MCF-7ALOX12 cells at 150 ?g/ml, MSCV Luciferase PGK-hygro plasmid and STAT3 Reporter HeLa Stable Cell Line (stably transfected with Luciferase gene under the control of STAT3 promoter) maintained in DMEM supplemented with 10% FBS. It is suitable for the selection of cells transfected with Escherichia coli hygromycin resistance gene (hyg or hph), as well as for the maintenance of the Hygromycin phenotype of resistant cells. Consistent with its more specific effect on translocation, hygromycin B exhibits monophasic inhibition of polypeptide synthesis, suggesting that it binds to a single site on the ribosome.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hygromycin B was used for hygro resistance effect in the following cells transfected 293T cells with mixture of plasmids at 160 μg/ml, MCF-7ALOX12 cells at 150 μg/ml, MSCV Luciferase PGK-hygro plasmid and STAT3 Reporter HeLa Stable Cell Line (stably transfected with Luciferase gene under the control of STAT3 promoter) maintained in DMEM supplemented with 10% FBS. It is suitable for the selection of cells transfected with Escherichia coli hygromycin resistance gene (hyg or hph), as well as for the maintenance of the Hygromycin phenotype of resistant cells. Consistent with its more specific effect on translocation, hygromycin B exhibits monophasic inhibition of polypeptide synthesis, suggesting that it binds to a single site on the ribosome.

Solubility
Fully miscible in water and also soluble in methanol or ethanol and buffer solution.

Notes
Protect from heat. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store in freezer at -20°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31282-04-9
Molecular Formula C20H37N3O13
Molecular Weight (g/mol) 527.524
MDL Number MFCD06795479
InChI Key GRRNUXAQVGOGFE-BBMONYMYSA-N
PubChem CID 134129613
IUPAC Name (3'S,3aS,4S,4'S,5'R,6R,6'R,7R,7aS)-4-[(1R,2S,3R,5S,6S)-3-amino-2,6-dihydroxy-5-(methylamino)cyclohexyl]oxy-6'-[(1S)-1-amino-2-hydroxyethyl]-6-(hydroxymethyl)spiro[4,6,7,7a-tetrahydro-3aH-[1,3]dioxolo[4,5-c]pyran-2,2'-oxane]-3',4',5',7-tetrol
SMILES CNC1CC(C(C(C1O)OC2C3C(C(C(O2)CO)O)OC4(O3)C(C(C(C(O4)C(CO)N)O)O)O)O)N

Specifications

Type Hygromycin B
Physical Form Liquid
Concentration 50mg/mL in distilled water
Quantity 5 x 1 mL
UN Number UN2810
Solubility Information Fully miscible in water and also soluble in methanol or ethanol and buffer solution.
Chemical Name or Material Hygromycin B, sterile-filtered
Hazard Category H301+H331-H310-H317-H318-H334
Hazard Statement GHS H Statement
H301-H330-H334-H318-H312-H317
Toxic if swallowed.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
Harmful in contact with skin.
May cause an allergic skin reaction.
Precautionary Statement P261-P262-P264b-P270-P271-P272-P280-P285-P301+P310-P302+P350-P304+P340-P305+P351+P338-P310-P311-P330-P333+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 250-545-5
RTECSNumber WK2130000
TSCA No
storageNote1 −30° to −10°C
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.