Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HYPE/HIP13 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981593
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1593 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1593 Supplier Bioss Supplier No. BS11698R
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The function remains known.

Specifications

Antigen HYPE/HIP13
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Ficd
Gene Accession No. Q9BVA6
Gene Alias adenosine monophosphate-protein transferase FICD; AMPylator FICD; BOS_16640; D5Ertd40e; De-AMPylase FICD; FIC domain containing; FIC domain-containing protein; fic S-phase protein cell division homolog; FICD; HIP13; HIP-13; huntingtin interacting protein 13; Huntingtin interacting protein E; huntingtin interactor protein E; Huntingtin yeast partner E; Huntingtin-interacting protein 13; Huntingtin-interacting protein E; HYPE; MGC5623; Protein adenylyltransferase FICD; UNQ3041; UNQ3041/PRO9857
Gene Symbols Ficd
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human HYPE.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11153
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.