Learn More
Epredia™ Hyperclean™ TruAir™ Ductless Fume Hood Filters
Catalog No. B1010011 Shop All Epredia Products
Change view
Click to view available options
Filter Type:
ACI Inorganic Acid
Charcoal GP
Formaldehyde
HEPA
Particle Pre-filter
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Filter Type
B1010011 Particle Pre-filter
B1010015 HEPA
B1010014 Formaldehyde
B1010013 ACI Inorganic Acid
B1010012 Charcoal GP
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. B1010011 Supplier Epredia™ Supplier No. B1010011
Add to Cart
Add to Cart
Add convenience and functionality to your fume hood with Epredia™ Hyperclean™ TruAir™ Ductless Fume Hood Accessories.
Specifications
Filter Type Particle Pre-filter
For Use With (Equipment) All
Quantity 6 Pk.
Product Type Ductless Fume Hood Accessories
Product Line TruAirHyperclean
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov