Learn More
Thermo Scientific™ HyPerforma™ 50 to 500L Tank Liners, 50L
Thermo Scientific™ HyPerforma™ 50 to 500L Tank Liners are available with or without bottom drain and are provided sterile for handling fluids in bioproduction applications.
Supplier: Thermo Scientific™ SH30647.01
Description
Thermo Scientific™ HyPerforma™ 50 to 500L Tank Liners are available with or without bottom drain and are provided sterile for handling fluids in bioproduction applications.
Tank Liner Key Features:
- Available in sizes that fit into industry standard cylindrical tanks and in Thermo Scientific HyPerforma Drums (SV3011X, SV50076, and SV50517 series)
- Gusset design allows for a close fit to the tank
- Chamber constructed of Thermo Scientific CX3-9 film, which provides the same contact surface as other Thermo Scientific BPCs
- Supplied gamma irradiated
- Customization options are available by contacting one of our Thermo Scientific Representatives today
Standard Tank Liners for your liquid applications available in-stock in the following configurations:
SH30647.01 to .03:
- 50 to 200L Tank Liner
- Support Container Needed: Design to fit into HyPerforma Flat Bottom PE Drums (SV50076 series)
- Quantity: Each = 1 Liner
SH30647.04 to .07:
- 50 to 500L Tank Liner
- Support Container Needed: Designed to fit into Industry Standard Cylindrical Tanks (SV3011X series)
- Quantity: Each = 1 Liner
SH30646 Series:
- 50 to 200L Tank Liner with One Bottom Drain
- Bottom Lineset 1: 72in. (182.9cm) of tubing leading to a Quick Disconnect (MPX series) Insert (male) connector
- Support Container Needed: Designed to fit into HyPerforma Conical Bottom PE Drums (SV50517 series)
Ordering Information:
Supplied double polybagged and irradiated with Certificate of Analysis.
Recommended for:
- Hydrations of powdered media and buffers
- Pooling of non-sterile solutions and fluids
- Non-aseptic collection of waste liquids from separation and purification systems
To order, use the part numbers listed in the above table.
Specifications
50 L | |
Tank Liner | |
Packed and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area. | |
Irradiated | |
CX3-9 |
Plastic | |
Packages are placed in strong cardboard cartons with product and lot identification. | |
SV50076.02 | |
HyPerforma™ | |
Sterile |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.