Learn More
Indofine Chemical Hyperoside , 98%, 50 Mg
Supplier: Indofine Chemical 06047 50 MG
HYPEROSIDE 50 MG, Herbal Standards, Hyperin, Quercetin-3-galactoside, C21H20O12, MW: 464.38, 0.98, 482-36-0, MFCD00016933
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.