Learn More
Apexbio Technology LLC HyperScript™ III First-Strand cDNA Synthesis SuperMix (with gDNA wiper) 50rxn
SDP
Supplier:  Apexbio Technology LLC K158450
Encompass_Preferred
HyperScript III First-Strand cDNA Synthesis SuperMix with gDNA wiper is a reagent kit for first-strand cDNA synthesis from RNA Its core enzyme HyperScript III Reverse Transcriptase is an engineered M-MLV RT with reduced RNase H activity enhanced thermal stability and stronger template affinity enabling improved reverse transcription The kit synthesizes cDNA from RNA templates includes a gDNA wiper reagent to remove genomic DNA and provides both Random Primers and Oligo(dT)23VN primers for flexible priming With high sensitivity ability to generate long cDNA (up to 12 3 kb) and tolerance to complex or GC-rich RNA it is well suited for RT-PCR and qPCR workflows
Catalog No. 50-000-97349
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.