Learn More
Thermo Scientific™ HyperSep Carboxylic Acid (WCX) SPE Cartridge
Description
Specifications
Specifications
| Column Capacity (Metric) | 6 mL |
| Bed Weight | 500 mg |
| Product Line | HyperSep |
| Type | SPE Cartridge |
| Quantity | 30/pk |