Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ HyperSep™ Polypropylene Lab Plates

Catalog No. 03250568
Change view
Click to view available options
Sorbent:
HyperSep C4
HyperSep C8
HyperSep Hypercarb
HyperSep Hypercarb and C18 (Mix Mode)
HyperSep SAX
HyperSep SCX
HyperSep Zirconium Dioxide
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sorbent
03-250-568 HyperSep Zirconium Dioxide
03-250-564 HyperSep C8
03-250-565 HyperSep C4
03-250-566 HyperSep Hypercarb
03-250-567 HyperSep Hypercarb and C18 (Mix Mode)
03-250-570 HyperSep SCX
03-250-571 HyperSep SAX
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 03-250-568 Supplier Thermo Scientific™ Supplier No. 60110306

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Get simple, convenient purification and sample preparation for proteins, DNA, RNA and other biomolecules with these 96-well lab plates.

Available in a range of chromatographic materials, Thermo Scientific™ HyperSep™ Polypropylene Lab Plates enable simple, convenient purification and sample preparation for proteins, DNA, RNA and other biomolecules. In this 96 well plate format, media is embedded at the bottom of the plate. Typical applications include:

  • Tissue culture and separation of products
  • Concentration of small-scale samples
  • Sample clean-up
  • Collection of sample after chromatography

The convenient 96-well HyperSep Lab Plates can be processed manually or using a liquid-handling robot. They are not suitable for use with a vacuum.
TRUSTED_SUSTAINABILITY

Specifications

Description Zirconium Dioxide; Polypropylene
For Use With (Application) Purification and Sample Preparation of Proteins, DNA, RNA and Other Biomolecules
Product Line HyperSep
Sorbent HyperSep Zirconium Dioxide
Type 96-Well Plate

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.