Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ HyperSep™ Polystyrene Lab Plates

Catalog No. 03250556
Change view
Click to view available options
Description:
C18 plus Hypercarb; Polystyrene
C4; Polystyrene
C8; Polystyrene
Hypercarb; Polystyrene
SCX; Polystyrene
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
03-250-556 C4; Polystyrene
03-250-555 C8; Polystyrene
03-250-557 Hypercarb; Polystyrene
03-250-558 C18 plus Hypercarb; Polystyrene
03-250-561 SCX; Polystyrene
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 03-250-556 Supplier Thermo Scientific™ Supplier No. 60110203

Get simple, convenient purification and sample preparation for proteins, DNA, RNA and other biomolecules with these 96-well lab plates.

Available in a range of chromatographic materials, Thermo Scientific™ HyperSep™ Polystyrene Lab Plates enable simple, convenient purification and sample preparation for proteins, DNA, RNA and other biomolecules. In this 96 well plate format, media is embedded at the bottom of the plate. Typical applications include:

  • Tissue culture and separation of products
  • Concentration of small-scale samples
  • Sample clean-up
  • Collection of sample after chromatography

The convenient 96-well HyperSep Lab Plates can be processed manually or using a liquid-handling robot. They are not suitable for use with a vacuum.
TRUSTED_SUSTAINABILITY

Specifications

Description C4; Polystyrene
For Use With (Application) Purification and Sample Preparation of Proteins, DNA, RNA and Other Biomolecules
Product Line HyperSep
Sorbent HyperSep C4
Type 96-Well Plate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.