Learn More
Thermo Scientific™ HyperSep™ Polystyrene Lab Plates
Description
Available in a range of chromatographic materials, Thermo Scientific™ HyperSep™ Polystyrene Lab Plates enable simple, convenient purification and sample preparation for proteins, DNA, RNA and other biomolecules. In this 96 well plate format, media is embedded at the bottom of the plate. Typical applications include:
- Tissue culture and separation of products
- Concentration of small-scale samples
- Sample clean-up
- Collection of sample after chromatography
Specifications
Specifications
| Description | Hypercarb; Polystyrene |
| For Use With (Application) | Purification and Sample Preparation of Proteins, DNA, RNA and Other Biomolecules |
| Particle Size | 30 μm |
| Product Line | HyperSep |
| Sorbent | HyperSep Hypercarb |
| Type | 96-Well Plate |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.