Learn More
Thermo Scientific™ MEPS™ for GC: Thermo Scientific, CTC Analytics, HTA and Varian 8400 Systems
Description
Save hours in sample preparation, and go from extraction to injection in a single process with Thermo Scientific™ HyperSep™ MEPS™ (Micro Extraction by Packed Sorbent) technology.
Suitable for reversed phases, normal phases, mixed mode or ion exchange chemistries, Thermo Scientific HyperSep MEPS Syringes and Components for Analytics are available in a variety of SPE phase chemistries. The HyperSep MEPS Barrel Insert and Needle Assembly (BIN) contains the stationary phase, and is built into the syringe needle.
These HyperSep MEPS BINs are designed for use with Thermo Scientific, CTC Analytics, HTA and Varian 8400 GC Systems.
Mechanically robust, the technique offers significant advantages over conventional SPE extraction for method development and other GC applications:
Significantly reduces the time to prepare and inject samples from hours to minutes—typically less than two minutes per sample
Eliminates all extra steps between sample preparation and sample injection; the technique is fully compatible with direct injection onto GC
Reduces buffer and solvent volume from milliliters to microliters
Reduces the required sample volume to as little as 3.6µL, making the techniques especially well-suited for on-line use with volume-limited samples
Can easily be automated compatible and is compatible with CTC and eVol platforms
Specifications
Specifications
| Description | Retain-AX MEPS for GC |
| For Use With (Application) | Gas Chromatography |
| For Use With (Equipment) | Thermo Scientific, CTC Analytics, HTA, and Varian 8400 Systems |
| Product Line | HyperSep |
| Sorbent | HyperSep Retain AX MEPS |
| Type | MEPS Barrel Insert and Needle (BIN) |