Learn More
Thermo Scientific™ MEPS™ for GC: Thermo Scientific, CTC Analytics, HTA and Varian 8400 Systems
Catalog No. 60308208
Change view
Click to view available options
Description:
C18 MEPS for GC
Hypercarb MEPS for GC
Retain PEP MEPS for GC
Retain PEP,-CX, -AX, Hypercarb, C18
Retain-AX MEPS for GC
Retain-CX MEPS for GC
Silica MEPS for GC
Verify-AX MEPS for GC
Verify-CX MEPS for GC
Sorbent:
HyperSep C18 MEPS
HyperSep Hypercarb MEPS
HyperSep Retain AX MEPS
HyperSep Retain CX MEPS
HyperSep Retain PEP MEPS
HyperSep Silica MEPS
HyperSep Verify AX MEPS
HyperSep Verify CX MEPS
Retain PEP, Retain CX, Retain AX, Hypercarb, and C18
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description Sorbent
60-308-208 Silica MEPS for GC HyperSep Silica MEPS
60-308-201 Retain PEP MEPS for GC HyperSep Retain PEP MEPS
60-308-202 Retain-CX MEPS for GC HyperSep Retain CX MEPS
60-308-203 Retain-AX MEPS for GC HyperSep Retain AX MEPS
60-308-204 Hypercarb MEPS for GC HyperSep Hypercarb MEPS
60-308-205 Verify-CX MEPS for GC HyperSep Verify CX MEPS
60-308-206 Verify-AX MEPS for GC HyperSep Verify AX MEPS
60-308-207 C18 MEPS for GC HyperSep C18 MEPS
60-308-209 Retain PEP,-CX, -AX, Hypercarb, C18 Retain PEP, Retain CX, Retain AX, Hypercarb, and C18
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 60-308-208 Supplier Thermo Scientific™ Supplier No. 60308208
Save hours in sample preparation, and go from extraction to injection in a single process with these 250μL syringes for GC applications using CTC Analytics.
Save hours in sample preparation, and go from extraction to injection in a single process with Thermo Scientific™ HyperSep™ MEPS™ (Micro Extraction by Packed Sorbent) technology.
Suitable for reversed phases, normal phases, mixed mode or ion exchange chemistries, Thermo Scientific HyperSep MEPS Syringes and Components for Analytics are available in a variety of SPE phase chemistries. The HyperSep MEPS Barrel Insert and Needle Assembly (BIN) contains the stationary phase, and is built into the syringe needle.
These HyperSep MEPS BINs are designed for use with Thermo Scientific, CTC Analytics, HTA and Varian 8400 GC Systems.
MEPS performs the same functions as SPE—the removal of interfering matrix components and the selective isolation and concentration of analytes—except that it miniaturizes conventional SPE packed bed devices from milliliter bed volumes to microliter volumes, and incorporates them right into the syringe.
Mechanically robust, the technique offers significant advantages over conventional SPE extraction for method development and other GC applications:
Significantly reduces the time to prepare and inject samples from hours to minutes—typically less than two minutes per sample
Eliminates all extra steps between sample preparation and sample injection; the technique is fully compatible with direct injection onto GC
Reduces buffer and solvent volume from milliliters to microliters
Reduces the required sample volume to as little as 3.6µL, making the techniques especially well-suited for on-line use with volume-limited samples
Can easily be automated compatible and is compatible with CTC and eVol platforms
TRUSTED_SUSTAINABILITY
Specifications
Description Silica MEPS for GC
For Use With (Application) Gas Chromatography
For Use With (Equipment) Thermo Scientific, CTC Analytics, HTA, and Varian 8400 Systems
Product Line HyperSep
Sorbent HyperSep Silica MEPS
Type MEPS Barrel Insert and Needle (BIN)