Learn More
Thermo Scientific™ HyperSep™ Retain PEP Plates
Description
Get balanced retention for polar and nonpolar analytes with Thermo Scientific™ HyperSep™ Retain PEP Plates, packed with a high capacity, high purity, highly porous polystyrene DVB material modified with urea functional groups. With chemistry pH stable 0 to 14, the of these cartridges in SPE sample preparation can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.
- [bullet] High, consistent recoveries
- [bullet] Fast and easy sample preparation and method development
- [bullet] Reproducible, reliable sample preparation in a traditional format
- [bullet] Available in a wide range of chemistries and bed weights
- [bullet] Drugs and metabolites in biological fluids
- [bullet] Peptides in serum, plasma or biological fluids
- [bullet] Environmental samples
Specifications
Specifications
| Description | Retain PEP 30 mg/1 mL 96 Removable Well Plate, 1 pk |
| Bed Weight | 30 mg |
| For Use With (Application) | Retention of Polar and Nonpolar Analytes |
| Particle Size | 30 to 50 μm |
| Product Line | HyperSep |
| Sorbent | HyperSep Urea-Modified Polystyrene DVB |
| Type | Solid Phase Extraction Well Plate |
| Volume (Metric) | 1 mL |
| Volume (Metric) Well | 1 mL |
| Quantity | 1 Pack |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.