Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ HyperSep™ Retain PEP Cartridges

Catalog No. 03251895
Change view
Click to view available options
Bed Weight:
30 mg
60 mg
100 mg
150 mg
200 mg
500 mg
600 mg
1000 mg
Column Capacity (Metric):
1 mL
3 mL
6 mL
25 mL
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Bed Weight Column Capacity (Metric)
03-251-895 600 mg 6 mL
03-150-633 30 mg 1 mL
03-150-635 60 mg 3 mL
03-150-636 200 mg 3 mL
03-150-637 500 mg 3 mL
03-150-638 500 mg 6 mL
60-107-207 100 mg 6 mL
60-107-208 60 mg 6 mL
60-107-211 150 mg 6 mL
03-151-617 200 mg 6 mL
03-151-605 1000 mg 25 mL
60-107-217 100 mg 3 mL
60-107-218 1000 mg 6 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Catalog No. 03-251-895 Supplier Thermo Scientific™ Supplier No. 60107225
Add to Cart
Add to Cart

Get balanced retention for polar and nonpolar analytes with Thermo Scientific™ HyperSep™ Retain PEP Cartridges.

Get balanced retention for polar and nonpolar analytes with Thermo Scientific™ HyperSep™ Retain PEP Cartridges, packed with a high capacity, high purity, highly porous polystyrene DVB material modified with urea functional groups. With chemistry pH stable 0 to 14, the of these cartridges in SPE sample preparation can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.

Features:
Ideal for large samples, Hypersep Retain cartridges conform to industry standard configurations and allow efficient routine processing by vacuum, positive pressure or centrifugation.
  • High, consistent recoveries
  • Fast and easy sample preparation and method development
  • Reproducible, reliable sample preparation in a traditional format
  • Available in a wide range of chemistries and bed weights
Applications:
  • Drugs and metabolites in biological fluids
  • Peptides in serum, plasma or biological fluids
  • Environmental samples
For Use With: The Thermo Scientific™ HyperSep-96™ System
TRUSTED_SUSTAINABILITY

Specifications

Column Capacity (Metric) 6 mL
Description 600 mg bed weight; 6 mL column volume
Bed Weight 600 mg
For Use With (Application) Retention of Polar and Nonpolar Analytes
Product Line HyperSep
Sorbent HyperSep Urea-Modified Polystyrene DVB
Type Solid Phase Extraction Cartridge
Volume (Metric) 6 mL

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.