Learn More
Thermo Scientific™ HyperSep™ Servo AMP Columns and Well Plates
Description
Thermo Scientific™ HyperSep Servo AMP Columns and Well Plates provide an optimized SPE solution for extraction of AMP and associated metabolites.
Greater performance helps provide improved confidence in results. HyperSep Servo+ and HyperSep Servo SPE product ranges remove lack of confidence in your sample preparation methodology by supplying an optimized extraction solution for analysis of Drugs of Abuse.
- Ready-to-use, optimized solutions
- Greater reproducibility and cleanliness of extract
- Greater confidence in your results
Specifications
Specifications
| Description | 300 mg/3 mL SPE Column 50 Pk |
| Bed Weight | 300 mg |
| For Use With (Application) | Analysis of Drugs of Abuse |
| Product Line | HyperSep |
| Sorbent | HyperSep Servo AMP |
| Type | Solid Phase Extraction Column |
| Volume (Metric) | 3 mL |