Learn More
Thermo Scientific™ HyperSep™ Servo Cocaine SPE Cartridges and Well Plates
For high confidence screening for drugs of abuse, these SPE cartridges and plates offer a robust, ready-to-use solution for extraction of cocaine class compounds.
Specifications
For Use With (Application) Analysis of Drugs of Abuse
Product Line HyperSep
Sorbent HyperSep Servo Cocaine
View More Specs
Products 10
Catalog Number Mfr. No. Description Price Quantity & Availability  
Catalog Number Mfr. No. Description Price Quantity & Availability  
60-111-701C
View Documents
Thermo Scientific™
60111701C
130 mg/1 mL SPE Column 100 Pk N/A N/A N/A
60-111-701P
View Documents
Thermo Scientific™
60111701P
96-well plate 10 mg 1 Pk N/A N/A N/A
60-111-702C
View Documents
Thermo Scientific™
60111702C
300 mg/3 mL SPE Column 50 Pk N/A N/A N/A
60-111-702P
View Documents
Thermo Scientific™
60111702P
96-well plate 25 mg 1 Pk N/A N/A N/A
60-111-703C
View Documents
Thermo Scientific™
60111703C
500 mg/3 mL SPE Column 50 Pk N/A N/A N/A
60-111-703P
View Documents
Thermo Scientific™
60111703P
96-well plate 50 mg 1 Pk N/A N/A N/A
60-111-704C
View Documents
Thermo Scientific™
60111704C
200 mg/6 mL SPE Column 50 Pk N/A N/A N/A
60-111-704P
View Documents
Thermo Scientific™
60111704P
96-well plate 100 mg 1 Pk N/A N/A N/A
60-111-705C
View Documents
Thermo Scientific™
60111705C
500 mg/6 mL SPE Column 30 Pk N/A N/A N/A
60-111-706C
View Documents
Thermo Scientific™
60111706C
1 g/6 mL SPE Column 30 Pk N/A N/A N/A
Description
Description
Get the cleanest possible sample extracts—and maximum confidence—when preparing samples for screening for drugs of abuse. Depending on your analytical needs, you can choose from a comprehensive range of Thermo Scientific™ HyperSep Servo products, each optimized for a different class of drugs. To ensure success, each product comes with detailed extraction procedures.
Thermo Scientific™ HyperSep Servo Cocaine Cartridges and Well Plates offer a robust, ready-to-use solution for dedicated extraction of cocaine class compounds.
HyperSep Servo and Servo+ Cartridges and Plates
Thermo Scientific™ HyperSep Servo and Servo+ cartridges and plates provide complete, high-reliability SPE sample preparation solutions for drug screening applications.
Both Servo and Servo+ product lines deliver the high levels of reproducibility and clean sample extracts required for maximum confidence when dealing with complex biological sample matrices. Servo+ cartridges and plates offer an extra measure of selectivity, higher loading capacities and enhanced robustness.
HyperSep SPE Cartridges and Plates
Thermo Scientific™ HyperSep solid phase extraction cartridges offer reproducible, reliable sample preparation in a traditional format. Available in a wide range of chemistries and bed weights, the cartridges are ideal for large samples and conform to industry standard configurations, allowing efficient routine processing by vacuum, by positive pressure or centrifugation.
The Thermo Scientific™ HyperSep-96™ system incorporates traditional solid phase extraction sorbents into 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customizable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing. HyperSep-96 products are either available in packs of 100 individual wells or in pre-assembled plates.
Specifications
Specifications
Analysis of Drugs of Abuse
HyperSep
HyperSep Servo Cocaine
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.