Learn More
Thermo Scientific™ HyperSep™ Servo+ PCP SPE Cartridges and Well Plates
For high confidence screening for drugs of abuse, these SPE cartridges and plates offer a robust, ready-to-use solution for extraction of PCP class compounds.
Specifications
For Use With (Application) Analysis of Drugs of Abuse
Product Line HyperSep
Sorbent HyperSep Servo+ PCP
View More Specs
Products 14
Catalog Number Mfr. No. Description Bed Weight Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Description Bed Weight Volume (Metric) Price Quantity & Availability  
60-110-601C
View Documents
Thermo Scientific™
60110601C
30 mg/1 mL SPE Column 100 Pk 30 mg 1 mL N/A N/A N/A
60-110-601P
View Documents
Thermo Scientific™
60110601P
96-well plate 5 mg 1 Pk 5 mg N/A N/A N/A
60-110-602C
View Documents
Thermo Scientific™
60110602C
30 mg/3 mL SPE Column 50 Pk 30 mg 3 mL N/A N/A N/A
60-110-602P
View Documents
Thermo Scientific™
60110602P
96-well plate 10 mg 1 Pk 10 mg N/A N/A N/A
60-110-603C
View Documents
Thermo Scientific™
60110603C
60 mg/3 mL SPE Column 50 Pk 60 mg 3 mL N/A N/A N/A
60-110-603P
View Documents
Thermo Scientific™
60110603P
96-well plate 30 mg 1 Pk 30 mg N/A N/A N/A
60-110-604C
View Documents
Thermo Scientific™
60110604C
200 mg/3 mL SPE Column 50 Pk 200 mg 3 mL N/A N/A N/A
60-110-605C
View Documents
Thermo Scientific™
60110605C
500 mg/3 mL SPE Column 50 Pk 500 mg 3 mL N/A N/A N/A
60-110-606C
View Documents
Thermo Scientific™
60110606C
500 mg 6 mL SPE Column 30 Pk 500 mg 6 mL N/A N/A N/A
60-110-607C
View Documents
Thermo Scientific™
60110607C
100 mg 6 mL SPE Column 30 Pk 100 mg 6 mL N/A N/A N/A
60-110-608C
View Documents
Thermo Scientific™
60110608C
60 mg 6 mL SPE Column 30 Pk 60 mg 6 mL N/A N/A N/A
60-110-609C
View Documents
Thermo Scientific™
60110609C
150 mg 6 mL SPE Column 30 Pk 150 mg 6 mL N/A N/A N/A
60-110-610C
View Documents
Thermo Scientific™
60110610C
200 mg 6 mL SPE Column 30 Pk 200 mg 6 mL N/A N/A N/A
60-110-611C
View Documents
Thermo Scientific™
60110611C
1 g 25 mL SPE Column 20 Pk 1 g 25 mL N/A N/A N/A
Description
Description
Get the cleanest possible sample extracts—and maximum confidence—when preparing samples for screening for drugs of abuse. Depending on your analytical needs, you can choose from a comprehensive range of Thermo Scientific™ HyperSep Servo+ products, each optimized for a different class of drugs. To ensure success, each product comes with detailed extraction procedures.
Thermo Scientific™ HyperSep Servo+ PCP Cartridges and Well Plates offer a robust, ready-to-use solution for dedicated extraction of PCP class compounds.
HyperSep Servo and Servo+ Cartridges and Plates
Thermo Scientific™ HyperSep Servo and Servo+ cartridges and plates provide complete, high-reliability SPE sample preparation solutions for drug screening applications.
Both Servo and Servo+ product lines deliver the high levels of reproducibility and clean sample extracts required for maximum confidence when dealing with complex biological sample matrices. Servo+ cartridges and plates offer an extra measure of selectivity, higher loading capacities and enhanced robustness.
HyperSep SPE Cartridges and Plates
Thermo Scientific™ HyperSep solid phase extraction cartridges offer reproducible, reliable sample preparation in a traditional format. Available in a wide range of chemistries and bed weights, the cartridges are ideal for large samples and conform to industry standard configurations, allowing efficient routine processing by vacuum, by positive pressure or centrifugation.
The Thermo Scientific™ HyperSep-96™ system incorporates traditional solid phase extraction sorbents into 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customizable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing. HyperSep-96 products are either available in packs of 100 individual wells or in pre-assembled plates.
Specifications
Specifications
Analysis of Drugs of Abuse
HyperSep
HyperSep Servo+ PCP
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.