Learn More
Thermo Scientific™ HyperSep™ Servo+ THC SPE Cartridges and Well Plates
For high confidence screening for drugs of abuse, these SPE cartridges and plates offer a robust, ready-to-use solution for extraction of THC class compounds.
Specifications
For Use With (Application) Analysis of Drugs of Abuse
Product Line HyperSep
Sorbent HyperSep Servo+ THC
View More Specs
Products 14
Catalog Number Mfr. No. Description Type Bed Weight Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Description Type Bed Weight Volume (Metric) Price Quantity & Availability  
60-110-401C
View Documents
Thermo Scientific™
60110401C
30 mg/1 mL SPE Column 100 Pk Solid Phase Extraction Column 30 mg 1 mL N/A N/A N/A
60-110-401P
View Documents
Thermo Scientific™
60110401P
96-well plate 5 mg 1 Pk 96-Well Plate 5 mg N/A N/A N/A
60-110-402C
View Documents
Thermo Scientific™
60110402C
30 mg/3 mL SPE Column 50 Pk Solid Phase Extraction Column 30 mg 3 mL N/A N/A N/A
60-110-402P
View Documents
Thermo Scientific™
60110402P
96-well plate 10 mg 1 Pk 96-Well Plate 10 mg N/A N/A N/A
60-110-403C
View Documents
Thermo Scientific™
60110403C
60 mg/3 mL SPE Column 50 Pk Solid Phase Extraction Column 60 mg 3 mL N/A N/A N/A
60-110-403P
View Documents
Thermo Scientific™
60110403P
96-well plate 30 mg 1 Pk 96-Well Plate 30 mg N/A N/A N/A
60-110-404C
View Documents
Thermo Scientific™
60110404C
200 mg/3 mL SPE Column 50 Pk Solid Phase Extraction Column 200 mg 3 mL N/A N/A N/A
60-110-405C
View Documents
Thermo Scientific™
60110405C
500 mg/3 mL SPE Column 50 Pk Solid Phase Extraction Column 500 mg 3 mL N/A N/A N/A
60-110-406C
View Documents
Thermo Scientific™
60110406C
500 mg 6 mL SPE Column 30 Pk Solid Phase Extraction Column 500 mg 6 mL N/A N/A N/A
60-110-407C
View Documents
Thermo Scientific™
60110407C
100 mg 6 mL SPE Column 30 Pk Solid Phase Extraction Column 100 mg 6 mL N/A N/A N/A
60-110-408C
View Documents
Thermo Scientific™
60110408C
60 mg 6 mL SPE Column 30 Pk Solid Phase Extraction Column 60 mg 6 mL N/A N/A N/A
60-110-409C
View Documents
Thermo Scientific™
60110409C
150 mg 6 mL SPE Column 30 Pk Solid Phase Extraction Column 150 mg 6 mL N/A N/A N/A
60-110-410C
View Documents
Thermo Scientific™
60110410C
200 mg 6 mL SPE Column30 Pk Solid Phase Extraction Column 200 mg 6 mL N/A N/A N/A
60-110-411C
View Documents
Thermo Scientific™
60110411C
1 g/25 mL SPE Column Solid Phase Extraction Column 1 g 25 mL N/A N/A N/A
Description
Description
Get the cleanest possible sample extracts—and maximum confidence—when preparing samples for screening for drugs of abuse. Depending on your analytical needs, you can choose from a comprehensive range of Thermo Scientific™ HyperSep Servo+ products, each optimized for a different class of drugs. To ensure success, each product comes with detailed extraction procedures.
Thermo Scientific™ HyperSep Servo+ THC Cartridges and Well Plates offer a robust, ready-to-use solution for dedicated extraction of THC and associated metabolites.
HyperSep Servo and Servo+ Cartridges and Plates
Thermo Scientific™ HyperSep Servo and Servo+ cartridges and plates provide complete, high-reliability SPE sample preparation solutions for drug screening applications.
Both Servo and Servo+ product lines deliver the high levels of reproducibility and clean sample extracts required for maximum confidence when dealing with complex biological sample matrices. Servo+ cartridges and plates offer an extra measure of selectivity, higher loading capacities and enhanced robustness.
HyperSep SPE Cartridges and Plates
Thermo Scientific™ HyperSep solid phase extraction cartridges offer reproducible, reliable sample preparation in a traditional format. Available in a wide range of chemistries and bed weights, the cartridges are ideal for large samples and conform to industry standard configurations, allowing efficient routine processing by vacuum, by positive pressure or centrifugation.
The Thermo Scientific™ HyperSep-96™ system incorporates traditional solid phase extraction sorbents into 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customizable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing. HyperSep-96 products are either available in packs of 100 individual wells or in pre-assembled plates.
Specifications
Specifications
Analysis of Drugs of Abuse
HyperSep
HyperSep Servo+ THC
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.