Learn More
Thermo Scientific™ HyperSep™ Servo Total-A SPE Cartridges and Well Plates
Screening for unknown acidic components when analyzing drugs of abuse? These SPE cartridges and plates offer a robust, ready-to-use solution.
Specifications
For Use With (Application) Analysis of Drugs of Abuse
Product Line HyperSep
Sorbent HyperSep Servo Total A
View More Specs
Products 10
Catalog Number Mfr. No. Description Price Quantity & Availability  
Catalog Number Mfr. No. Description Price Quantity & Availability  
60111101C
View Documents
Thermo Scientific™
60111101C
130 mg/1 mL SPE Column 100 Pk N/A N/A N/A
60111101P
View Documents
Thermo Scientific™
60111101P
96-well plate 5 mg 1 Pk N/A N/A N/A
60111102C
View Documents
Thermo Scientific™
60111102C
300 mg/3 mL SPE Column 50 Pk N/A N/A N/A
60111102P
View Documents
Thermo Scientific™
60111102P
96-well plate 10 mg 1 Pk N/A N/A N/A
60111103C
View Documents
Thermo Scientific™
60111103C
500 mg/3 mL SPE Column 50 Pk N/A N/A N/A
60111103P
View Documents
Thermo Scientific™
60111103P
96-well plate 30 mg 1 Pk N/A N/A N/A
60111104C
View Documents
Thermo Scientific™
60111104C
200 mg/6 mL SPE Column 50 Pk N/A N/A N/A
60111104P
View Documents
Thermo Scientific™
60111104P
96-well plate 100 mg 1 Pk N/A N/A N/A
60111105C
View Documents
Thermo Scientific™
60111105C
500 mg/6 mL SPE Column 30 Pk N/A N/A N/A
60111106C
View Documents
Thermo Scientific™
60111106C
1 g/6 mL SPE Column 30 Pk N/A N/A N/A
Description
Description
Get the cleanest possible sample extracts—and maximum confidence—when preparing samples for screening for drugs of abuse. Depending on your analytical needs, you can choose from a comprehensive range of Thermo Scientific™ HyperSep Servo products, each optimized for a different class of drugs. To ensure success, each product comes with detailed extraction procedures.
Thermo Scientific™ HyperSep Servo Total-A Cartridges and Well Plates offer a robust, ready-to-use solution for screening of a wide range of unknown acidic compounds.
HyperSep Servo and Servo+ Cartridges and Plates
Thermo Scientific™ HyperSep Servo and Servo+ cartridges and plates provide complete, high-reliability SPE sample preparation solutions for drug screening applications.
Both Servo and Servo+ product lines deliver the high levels of reproducibility and clean sample extracts required for maximum confidence when dealing with complex biological sample matrices. Servo cartridges and plates offer an extra measure of selectivity, higher loading capacities and enhanced robustness.
HyperSep SPE Cartridges and Plates
Thermo Scientific™ HyperSep solid phase extraction cartridges offer reproducible, reliable sample preparation in a traditional format. Available in a wide range of chemistries and bed weights, the cartridges are ideal for large samples and conform to industry standard configurations, allowing efficient routine processing by vacuum, by positive pressure or centrifugation.
The Thermo Scientific™ HyperSep-96™ system incorporates traditional solid phase extraction sorbents into 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customizable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing. HyperSep-96 products are either available in packs of 100 individual wells or in pre-assembled plates.
Specifications
Specifications
Analysis of Drugs of Abuse
HyperSep
HyperSep Servo Total A
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.