Learn More
Thermo Scientific™ HyperSep™ Solid Phase Extraction (SPE) Accessories, Empty Wells
Catalog No. 03052230
Change view
Click to view available options
Type:
Empty Fritted Well
Empty Wells
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
03-052-230 Empty Fritted Well
03-052-231 Empty Wells
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 03-052-230 Supplier Thermo Scientific™ Supplier No. 60300311
These empty 1mL wells are used with Thermo Scientific™ HyperSep 96-well plate applications.
The use of SPE sample preparation can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.
Thermo Scientific™ HyperSep Empty 1mL Wells are used in HyperSep 96-well plate applications. They are available in fritted or fritless configurations, in packs of 100.
HyperSep SPE Cartridges and Plates
HyperSep solid phase extraction cartridges offer reproducible, reliable sample preparation in a traditional format. Available in a wide range of chemistries and bed weights, the cartridges are ideal for large samples and conform to industry standard configurations, allowing efficient routine processing by vacuum, by positive pressure or centrifugation.
The HyperSep-96 system incorporates traditional solid phase extraction sorbents into 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customizable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing. HyperSep-96 products are available in packs of 100 individual wells and in pre-assembled plates.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) HyperSep-96 Products and Compatible Manifolds
Type Empty Fritted Well