Learn More
Thermo Scientific™ HyperSep™ Solid Phase Extraction (SPE) Sample Collection Plates
Description
Use Thermo Scientific™ HyperSep™ Solid Phase Extraction (SPE) Sample Collection Plates with your HyperSep system.
Fast analysis without sacrificing accuracy or reproducibility. Incorporate traditional solid phase extraction sorbents into a 96-well plate format using the HyperSep system.
- Innovative design of 96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation
- Well plates are customizable to allow any arrangement of wells with differing sorbent chemistries for simultaneous processing.
Ordering Information:
Available in packs of 100 individual wells or in pre-assembled plates.
Specifications
Specifications
| Description | Sample Collection Plate, 1 mL |
| For Use With (Application) | SPE Sample Preparation |
| For Use With (Equipment) | Vacuum Manifold |
| Product Line | HyperSep |
| Type | Sample Collection Plate |
| Volume (Metric) | 1 mL |