Learn More
Description
- Enhanced cleaning applications over a standard sodium hypochlorite solution
- Increased cleaning surface compatibility
- Designed for all washable non-porous environmental surfaces
Specifications
Specifications
| Product Type | Cleaner |
| Sterility | Sterile |
| Concentration | 0.52% |
| Format | SimpleMix |
| Includes | Attached Activator |
| Volume (English) | 1 gal. |
| Volume (Metric) | 3.8 L |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.