Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hypotaurine, 100.4%, MP Biomedicals™

Catalog No. ICN15971180 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15971180 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15971180 Supplier MP Biomedicals, Inc Supplier No. 0215971180

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 300-84-5
Molecular Formula C2H7NO2S
Molecular Weight (g/mol) 109.143
InChI Key VVIUBCNYACGLLV-UHFFFAOYSA-N
Synonym hypotaurine, ethanesulfinic acid, 2-amino, 2-aminoethylsulfinic acid, cystaminesulfinic acid, cystaminesulfinate, 2-aminoethylsulfinate, tocris-0208, 2-amino-ethanesulfinate, lopac-h-1384, acmc-1cmf0
PubChem CID 107812
ChEBI CHEBI:16668
IUPAC Name 2-aminoethanesulfinic acid
SMILES C(CS(=O)O)N

Specifications

Color White
Assay Percent Range 1.004
Quantity 100 mg
Formula Weight 109.15
Physical Form Prills
Chemical Name or Material Hypotaurine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.