accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2SOO

CAS RN 300-84-5

IUPAC Name: 2-aminoethane-1-sulfinate
Synonyms: 2-aminoethanesulfinate
Molecular Weight (g/mol): 108.14
Molecular Formula: C2H6NO2S
InChi Key: VVIUBCNYACGLLV-UHFFFAOYSA-M
SMILES: NCCS([O-])=O
Molecular Weight (g/mol) 
  • (1)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)
  • (1)