Learn More
Description
- Suitable for flow cytometry and fluorescence imaging.
- Spectrally compatible with FITC and R-PE.
Specifications
Specifications
| Quantity | 500 μL |
| Product Type | Fluorescent Dye |
| Content And Storage | Store at 4°C in the dark. Do not freeze. |
| Target | HypoxiTRAK(TM) |
| Label Type | Other Labels or Dyes |
| Product Line | Novus |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
