Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hypromellose, Substitution Type 2208, 100,000 mPa.s, USP, 4-12%, Spectrum™ Chemical
SDP

Catalog No. 18603384 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
Packaging:
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-384 500 g Poly Bottle
18-603-383 125 g Poly Bottle
18-603-385 2.5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-603-384 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. H1136500GM
Add to Cart
Add to Cart

H1136, 9004-65-3

Spectrum™ Chemical Hypromellose, Substitution Type 2208, 100,000 mPa.s, USP is a semisynthetic, viscoelastic polymer that is inert and used as an ophthalmic lubricant, a controlled-delivery component and an excipient in oral medicaments. It is also used as a food additive and acts as an emulsifier, suspending agent and thickening agent. Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 9004-65-3
Molecular Formula C56H108O30
Molecular Weight (g/mol) 1261.45
InChI Key PUSNGFYSTWMJSK-UHFFFAOYNA-N
IUPAC Name 1-{[3,4,5-tris(2-hydroxypropoxy)-6-{[4,5,6-tris(2-hydroxypropoxy)-2-[(2-hydroxypropoxy)methyl]oxan-3-yl]oxy}oxan-2-yl]methoxy}propan-2-ol; 3,4,5-trimethoxy-2-(methoxymethyl)-6-{[4,5,6-trimethoxy-2-(methoxymethyl)oxan-3-yl]oxy}oxane
SMILES COCC1OC(OC2C(COC)OC(OC)C(OC)C2OC)C(OC)C(OC)C1OC.CC(O)COCC1OC(OC2C(COCC(C)O)OC(OCC(C)O)C(OCC(C)O)C2OCC(C)O)C(OCC(C)O)C(OCC(C)O)C1OCC(C)O

Specifications

CAS Max % 100%
Quantity 500 g
Percent Purity 4 to 12%
Grade USP
Ignition Residue 1.5%
Loss on Drying 5%
Packaging Poly Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.