Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

I-Ak Mouse, anti-Mouse, R718, Clone: 11-5.2, BD OptiBuild™

Catalog No. BDB752251 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752251 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB752251 Supplier BD Biosciences Supplier No. 752251
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 11-5.2 monoclonal antibody specifically recognizes the α chain of the I-A[k] MHC class II alloantigen. It cross-reacts with cells from mice of the H-2[r] haplotype. mAb 11-5.2 recognizes the Ia.19 determinant, and it's reactivity is dependent upon the presence of Glu at position 75 of the Aα chain. Reactivity with other H-2 haplotypes (e.g., b, d, f, p, q, s) has not been observed.

TRUSTED_SUSTAINABILITY

Specifications

Antigen I-Ak
Applications Flow Cytometry
Classification Monoclonal
Clone 11-5.2
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.