Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Bordetella pertussis toxin IgG ELISA Kit

Catalog No. 89032292 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-032-292 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-032-292 Supplier Abnova™ Supplier No. KA4838
Add to Cart
Add to Cart

Enzyme-linked immunosorbent assay for the detection of toxin IgG antibodies to Bordetella pertussis.

Intended for detection of specific IgG antibodies in a sample by means of a sandwich type of the EIA method (i.e. a solid phase coated with specific antigen – antibody from the analyzed sample – labeled antibody).

Specifications

Description Bordetella pertussis toxin IgG ELISA Kit is an enzyme-linked immunosorbent assay for the detection of toxin IgG antibodies to Bordetella pertussis.
Conjugate HRP
Detection Method Colorimetric
Product Type Bordetella pertussis toxin IgG ELISA Kit
Sample Type Human Plasma, Human Serum
Target Species Human
For Use With (Application) Quantification
Kit Contents Microtitre Plate: 96 wells, Negative Control (Calibrator 1): 1 U/mL (2mL), Calibrator 2: 40 U/mL (3mL), Calibrator 3: 100 IU/mL (2mL), Calibrator 4 (Positive Control): 350 U/mL (2mL), Conjugate: 15mL, Sample Diluent 2: 105mL, TMB-Complete 2: 15mL, Wash Solution: 20x concentrated buffer: 75mL, Stop Solution: Acid solution: 15mL
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Volume 100 μL
Storage Requirements Store the kit at 4°C. Do not freeze.
Target Human
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.