Learn More
Selleck Chemical LLC I-BRD9 5mg 1714146-59-4 GSK602
SDP
Supplier:  Selleck Chemical LLC S78355MG
Encompass_Preferred
I-BRD9 (GSK602) is a potent and selective BRD9 inhibitor with pIC50 of 7.3, while it displayed a pIC50 of 5.3 against BRD4. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5320
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.