Learn More
TARGETMOL CHEMICALS INC I-CBP112 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T3969
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. I-CBP112 is a specific and potent acetyl-lysine competitive protein-protein interaction inhibitor targeting the CBP/p300 bromodomains. purity: 99%
Catalog No. 50-258-0040
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.