Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Enterobacter cloacae subsp. cloacae ATCC™ 13047™†, Microbiologics™
SDP

Catalog No. 23016134 Shop All Microbiologics Products
Change view
Click to view available options
Media Type:
Kwik-stik™
Lyfo disk™
Quantity:
2/Pk.
6/Pk.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Media Type Quantity
23-016-134 Kwik-stik™ 6/Pk.
23-016-584 Lyfo disk™ 6/Pk.
23-021077 Kwik-stik™ 2/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 23-016-134 Supplier MicroBioLogics Supplier No. 0323K
Add to Cart
Add to Cart
This item is not returnable. View return policy

For culture purposes and QC applications

KWIK-STIK™

  • Clever design and accurate, repeatable results
  • Each device features a single Enterobacter cloacae subsp. cloacae strain in a lyophilized pellet, a reservoir of hydrating fluid and inoculating swab
  • For added convenience, a peel-off identification label is included for easy documentation
  • Available in packs of 2 or 6
KWIK-STIK™ Plus
  • Five self-contained KWIK-STIK units with microorganism limited to two passages from reference culture
  • LYFO DISK™
    • The most economical reference stock culture option; lyophilized pellet containing a singleEnterobacter cloacae subsp. cloacae strain
    • Easy to use; simply re-hydrate the pellet and inoculate
    • Available in a re-sealable vial containing 6 pellets
    • ATCC™ 13047™†

    †The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.

    Specifications

    ATCC® ATCC 13047
    Biosafety Level 1
    Certifications/Compliance CE, online certificate of analysis
    Content And Storage 2°C to 8°C
    Form Lyophilized Pellet
    Media Type Kwik-stik™
    Product Type Quality Control Organism
    For Use With (Application) Culture purpose and QC application
    Quantity 6/Pk.
    Species Enterobacter cloacae subsp. cloacae
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.