Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mirus Bio™ flashBAC™ Baculovirus Expression System

Catalog No. MIR6115 Shop All Mirus Bio LLC Products
Change view
Click to view available options
No. of Reactions:
5
24
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
MIR6115 5
MIR6120 24
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MIR6115 Supplier Mirus Bio™ Supplier No. MIR 6115
Only null left
Add to Cart
Add to Cart

flashBAC™ Baculovirus Expression System is a streamlined platform for the production of recombinant baculoviruses. Optimal for expression of most nuclear or cytoplasmic proteins.

The original flashBAC expression vector. flashBAC expression systems are a streamlined platform for the production of recombinant baculoviruses. Genetic optimization of the Autographa californica nucleopolyhedrovirus (AcMNPV) genome yields recombinant virus in a single step and eliminates the need for separation of recombinant and parental virus by plaque purification. Protein expression and secretion is improved by deletion of the chitinase gene from the AcMNPV genome. Optimal for expression of most nuclear or cytoplasmic proteins.
  • Single-step baculovirus generation in insect cells, no plaque purification required
  • Ready to use with most transfer vectors, including BacMam
  • High recombinant protein yields

Warranty and Services

1 Year

Specifications

Content And Storage Store flashBAC™ DNA at 4°C and control plasmid at -20°C
Cell Type Insect Cells
Format Kit
Sample Type Baculovirus DNA, Plasmid DNA
Promoter None (Promoterless)
Transfection Technique Chemical Transfection
For Use With (Application) Transfection
No. of Reactions 5
Product Type Transfection Reagent
Sterility Sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.