Learn More
Description
- Single-step baculovirus generation in insect cells, no plaque purification required
- Ready to use with most transfer vectors, including BacMam
- High recombinant protein yields
Warranty and Services
1 Year
Specifications
Specifications
| Content And Storage | Store flashBAC™ DNA at 4°C and control plasmid at -20°C |
| Cell Type | Insect Cells |
| Format | Kit |
| Sample Type | Baculovirus DNA, Plasmid DNA |
| Promoter | None (Promoterless) |
| Transfection Technique | Chemical Transfection |
| For Use With (Application) | Transfection |
| No. of Reactions | 24 |
| Product Type | Transfection Reagent |
| Sterility | Sterile |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.