Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mirus Bio™ flashBAC™ ULTRA Baculovirus Expression System

Catalog No. MIR6140 Shop All Mirus Bio LLC Products
Change view
Click to view available options
No. of Reactions:
5
24
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
MIR6140 24
MIR6135 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MIR6140 Supplier Mirus Bio™ Supplier No. MIR 6140
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

flashBAC™ ULTRA Baculovirus Expression System is a streamlined platform for the production of recombinant baculoviruses. Ideal for the most difficult to express proteins including nuclear, cytoplasmic, membrane and secreted proteins.

flashBAC expression systems are a streamlined platform for the production of recombinant baculoviruses. Genetic optimization of the Autographa californica nucleopolyhedrovirus (AcMNPV) genome yields recombinant virus in a single step and eliminates the need for separation of recombinant and parental virus by plaque purification. flashBAC ULTRA is the most advanced flashBAC vector system, optimized by deletions in the chitinase, cathepsin, p10, p74 and p26 genes. The ideal system for the most difficult to express proteins including nuclear, cytoplasmic, membrane and secreted proteins.
  • Single-step baculovirus generation in insect cells, no plaque purification required
  • Ready to use with most transfer vectors, including BacMam
  • High recombinant protein yields

Warranty and Services

1 Year

Specifications

Content And Storage Store flashBAC™ DNA at 4°C and control plasmid at -20°C
Cell Type Insect Cells
Format Kit
Sample Type Baculovirus DNA, Plasmid DNA
Promoter None (Promoterless)
Transfection Technique Chemical Transfection
For Use With (Application) Transfection
No. of Reactions 24
Product Type Expression System
Sterility Sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.