Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mirus Bio™ Label IT™ Nucleic Acid Labeling Kit, Cy3

Catalog No. MIR3625 Shop All Mirus Bio LLC Products
Change view
Click to view available options
Quantity:
For 100 μg nucleic acid
For 5 μg nucleic acid
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MIR3625 For 5 μg nucleic acid
MIR3600 For 100 μg nucleic acid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MIR3625 Supplier Mirus Bio™ Supplier No. MIR 3625
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Label IT™ Reagent, Label IT™ reconstitution solution, 10X Labeling Buffer and solutions

Use the Label IT™ Cy3 Labeling Kit to efficiently and reproducibly attach marker molecules to DNA or RNA in a simple one-step reaction within minutes. Covalently attach labels without impacting hybridization performance.

  • Kits label any DNA or RNA template and are suitable for a wide range of applications
  • One-step chemical method provides easy and consistent control of the labeling reactions
  • Adjustable labeling density permits DNA and RNA to be optimally labeled for high sensitivity results
  • Covalent mechanism provides permanent, nondestructive modification of nucleic acid residues
  • Labels do not impact hybridization performance

Specifications

Detection Method Fluorescence
Format Kit
Label or Dye Cyanine3
Final Product Type DNA (Labeled), RNA (Labeled)
Labeling Target DNA (General), Oligos, RNA (General)
For Use With (Application) Efficient, direct, nonenzymatic labeling of DNA and RNA
Includes Label IT™ Reagent, Label IT™ reconstitution solution, 10X Labeling Buffer and solutions
Product Type Label IT Cy3 Labeling Kit
Quantity For 5 μg nucleic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.