Learn More
ZeptoMetrix Listeria monocytogenes serotype 1/2b, Genomic DNA
Shop All ZeptoMetrix Corporation Products

Description
- Can also be used to determine a limit of detection (LOD), in diagnostic assay development, cross-reactivity studies or genomic sequencing
- Each aliquot contains 10μg of DNA extracted from pure culture
- Supplied in TE Buffer
Controls should be run using the same protocols as those used to amplify extracted clinical specimens.
Amplification/detection control for nucleic acid testing of Listeria monocytogenes
Specifications
Specifications
| Certifications/Compliance | ISO, 13485 |
| Content And Storage | -20°C |
| Form | Solution |
| Product Type | Genomic DNA Bacteria Control |
| For Use With (Application) | Amplification/detection control for Nucleic Acid testing of Listeria monocytogenes |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.