Learn More
IKA™ I-MAG BR Stand Rod Holder
SDP
Catalog No. 01184872 Shop All IKA Works Products
Change view
Click to view available options
Type:
Stand Rod Holder
Catalog No. Type
01-184-872 Stand Rod Holder
1 options
This item is not returnable. View return policy
Catalog No. 01-184-872 Supplier IKA™ Supplier No. 20110383
Add to Cart
Add to Cart
This item is not returnable. View return policy
Stand rod holder for attachment to the bottom of the I-MAG industrial magnetic stirrer.
  • In combination with support rod H 16 V and strap clamp RH 3, vessels can be securely fastened to the magnetic stirrer and secured against rotation.
Specifications
Type Stand Rod Holder
Weight (Metric) 0.17 kg
Material Aluminum Alloy
Weight (English) 0.38 lb.