Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Phosphoric Acid, 85.0% W/W, MilliporeSigma™

Catalog No. 50948031 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Glass Bottle, Pre-Packed
Plastic Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
50-948-031 500 mL Glass Bottle, Pre-Packed
50-948-028 4 L Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-948-031 Supplier MilliporeSigma Supplier No. PX0995P5
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7664-38-2
Molecular Formula H3O4P
Molecular Weight (g/mol) 97.99
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
IUPAC Name phosphoric acid
SMILES OP(O)(O)=O

Specifications

pH < 0.5 (100 g/L, H2O, 20°C)
Melting Point 21°C
Boiling Point 158°C (1013 mbar)
Packaging Glass Bottle, Pre-Packed
Vapor Pressure 2 hPa (20°C)
Assay Percent Range 85.0% min
Quantity 500 mL
Recommended Storage Store at room temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.