Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Xylene, 98.0%, MilliporeSigma™

Catalog No. 50947524 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
3 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-947-524 3 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-947-524 Supplier MilliporeSigma Supplier No. XX00451

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 106-42-3
Molecular Weight (g/mol) 106.17

Specifications

Melting Point 13.3°C
Quantity 3 kg
Flash Point 24°C
Packaging Glass Bottle (Pre-packed)
Assay Percent Range 98.0% min
Vapor Pressure 8.7 hPa (20°C)
Solubility Information 0.2 g/L
Boiling Point 138.4°C (1013 hPa)
Recommended Storage Store at room temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.