CAS RN 106-42-3
p-Xylene, 99%
CAS: 106-42-3 Molecular Formula: C8H10 Molecular Weight (g/mol): 106.17 MDL Number: MFCD00008556 InChI Key: URLKBWYHVLBVBO-UHFFFAOYSA-N Synonym: p-xylene,para-xylene,1,4-dimethylbenzene,p-methyltoluene,p-dimethylbenzene,p-xylol,benzene, 1,4-dimethyl,4-xylene,chromar,4-methyltoluene PubChem CID: 7809 ChEBI: CHEBI:27417 IUPAC Name: 1,4-xylene SMILES: CC1=CC=C(C)C=C1
p-Xylene, 99+%, extra pure
CAS: 106-42-3 Molecular Formula: C8H10 Molecular Weight (g/mol): 106.17 MDL Number: MFCD00008556 InChI Key: URLKBWYHVLBVBO-UHFFFAOYSA-N Synonym: p-xylene,para-xylene,1,4-dimethylbenzene,p-methyltoluene,p-dimethylbenzene,p-xylol,benzene, 1,4-dimethyl,4-xylene,chromar,4-methyltoluene PubChem CID: 7809 ChEBI: CHEBI:27417 IUPAC Name: 1,4-xylene SMILES: CC1=CC=C(C)C=C1
p-Xylene, 99%, for HPLC
CAS: 106-42-3 Molecular Formula: C8H10 Molecular Weight (g/mol): 106.17 MDL Number: MFCD00008556 InChI Key: URLKBWYHVLBVBO-UHFFFAOYSA-N Synonym: p-xylene,para-xylene,1,4-dimethylbenzene,p-methyltoluene,p-dimethylbenzene,p-xylol,benzene, 1,4-dimethyl,4-xylene,chromar,4-methyltoluene PubChem CID: 7809 ChEBI: CHEBI:27417 IUPAC Name: 1,4-xylene SMILES: CC1=CC=C(C)C=C1
p-Xylene 99.0+%, TCI America™
CAS: 106-42-3 Molecular Formula: C8H10 Molecular Weight (g/mol): 106.17 MDL Number: MFCD00008556 InChI Key: URLKBWYHVLBVBO-UHFFFAOYSA-N Synonym: 1,4-Dimethylbenzene PubChem CID: 7809 ChEBI: CHEBI:27417 IUPAC Name: 1,4-xylene SMILES: CC1=CC=C(C)C=C1
p-Xylene, 99%, pure
CAS: 106-42-3 Molecular Formula: C8H10 Molecular Weight (g/mol): 106.17 MDL Number: MFCD00008556 InChI Key: URLKBWYHVLBVBO-UHFFFAOYSA-N Synonym: p-xylene,para-xylene,1,4-dimethylbenzene,p-methyltoluene,p-dimethylbenzene,p-xylol,benzene, 1,4-dimethyl,4-xylene,chromar,4-methyltoluene PubChem CID: 7809 ChEBI: CHEBI:27417 IUPAC Name: 1,4-xylene SMILES: CC1=CC=C(C)C=C1
p-Xylene, 99%, for spectroscopy
CAS: 106-42-3 Molecular Formula: C8H10 Molecular Weight (g/mol): 106.17 MDL Number: MFCD00008556 InChI Key: URLKBWYHVLBVBO-UHFFFAOYSA-N Synonym: 1, 4-Dimethylbenzene PubChem CID: 7809 ChEBI: CHEBI:27417 IUPAC Name: 1,4-xylene SMILES: CC1=CC=C(C)C=C1
p-Xylene, Pharmaceutical Secondary Standard, Certified Reference Material, MilliporeSigma™ Supelco™
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
p-Xylene, ≥99% (GC), Solstice
CAS: 106-42-3 Molecular Formula: C8H10 Molecular Weight (g/mol): 106.17 MDL Number: MFCD00008556 InChI Key: URLKBWYHVLBVBO-UHFFFAOYSA-N Synonym: p-xylene,para-xylene,1,4-dimethylbenzene,p-methyltoluene,p-dimethylbenzene,p-xylol,benzene, 1,4-dimethyl,4-xylene,chromar,4-methyltoluene PubChem CID: 7809 ChEBI: CHEBI:27417 IUPAC Name: 1,4-xylene SMILES: CC1=CC=C(C)C=C1
p-Xylene, Analytical Standard, ≥99.5% (GC), MilliporeSigma™ Supelco™
p-Xylene is an aromatic liquid, which is an isomeric form of xylene. It can be produced via Diels-Alder reaction between 2,5-dimethylfuran and ethylene catalyzed by zeolite.
BTEX Standard, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
p-Xylene 99.0+%, TCI America™
CAS: 106-42-3 Molecular Formula: C8H10 Molecular Weight (g/mol): 106.17 MDL Number: MFCD00008556 InChI Key: URLKBWYHVLBVBO-UHFFFAOYSA-N Synonym: p-xylene,para-xylene,1,4-dimethylbenzene,p-methyltoluene,p-dimethylbenzene,p-xylol,benzene, 1,4-dimethyl,4-xylene,chromar,4-methyltoluene PubChem CID: 7809 ChEBI: CHEBI:27417 IUPAC Name: 1,4-xylene SMILES: CC1=CC=C(C)C=C1
p-Xylene, SPEX CertiPrep™
CAS: 106-42-3 Molecular Formula: C8H10 Molecular Weight (g/mol): 106.17 MDL Number: MFCD00008556 InChI Key: URLKBWYHVLBVBO-UHFFFAOYSA-N PubChem CID: 7809 ChEBI: CHEBI:27417 IUPAC Name: 1,4-xylene SMILES: CC1=CC=C(C)C=C1
p-Xylene (Certified), Fisher Chemical™
CAS: 106-42-3 Molecular Formula: C8H10 Molecular Weight (g/mol): 106.17 MDL Number: MFCD00008556 InChI Key: URLKBWYHVLBVBO-UHFFFAOYSA-N Synonym: p-xylene,para-xylene,1,4-dimethylbenzene,p-methyltoluene,p-dimethylbenzene,p-xylol,benzene, 1,4-dimethyl,4-xylene,chromar,4-methyltoluene PubChem CID: 7809 ChEBI: CHEBI:27417 IUPAC Name: 1,4-xylene SMILES: CC1=CC=C(C)C=C1
p-Xylene, 98.0%, MilliporeSigma™
CAS: 106-42-3 Molecular Weight (g/mol): 106.17
p-Xylene in Dimethyl Sulfoxide, SPEX CertiPrep™
CAS: 106-42-3 Molecular Formula: C8H10 Molecular Weight (g/mol): 106.17 MDL Number: MFCD00008556 InChI Key: URLKBWYHVLBVBO-UHFFFAOYSA-N PubChem CID: 7809 ChEBI: CHEBI:27417 IUPAC Name: 1,4-xylene SMILES: CC1=CC=C(C)C=C1