Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medique I-Prin Tablets

Catalog No. 19090786 Shop All Medique Products
Change view
Click to view available options
Quantity:
2/Pk., 100/Box
2/Pk., 250/Box
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-090-786 2/Pk., 100/Box
19-090-785 2/Pk., 250/Box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-090-786 Supplier Medique Supplier No. 10047
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For relief of fever, menstrual cramps, arthritis, cold, etc.

  • Easy to swallow
  • Film coated
  • Each Tablet contains Ibuprofen 200mg

Specifications

Product Type Ibuprofen
Active Ingredients Ibuprofen (200 mg)
For Use With (Application) For relief from fever, menstrual cramps, arthritis, colds, headache, toothache, backache
Color White
Quantity 2/Pk., 100/Box
Recommended Age 12 years and older
Storage Requirements 68°F to 77°F (20°C to 25°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.