Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Silencer™ Cy™3-labeled GAPDH siRNA (human, mouse, rat)

Catalog No. AM4649
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 nmol
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM4649 5 nmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM4649 Supplier Invitrogen™ Supplier No. AM4649
Season of Savings
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Silencer™ Cy™3-labeled GAPDH siRNA (human, mouse, rat)

Invitrogen™ fluorescently labeled control siRNA for the easy monitoring of siRNA uptake by fluorescence microscopy along with siRNA-induced knockdown. Purified, annealed, and ready to use, the 5 nmol aliquot is supplied in a stable, dried format.

  • Fluorescently labeled siRNAs simplify the monitoring of siRNA delivery
  • Ideal for correlating siRNA uptake with mRNA knockdown
  • Functionally tested in several common cell lines
  • HPLC purified, duplexed, and ready to use

    Visualize siRNA Uptake with Labeled siRNAs
    Fluorescent labeling of siRNAs allows direct observation of their cellular uptake, distribution, and localization. Fluorescently labeled siRNAs are commonly used to monitor transfection efficiency during optimization of transfection conditions. In general, siRNAs are rapidly taken up by and distributed throughout cells in as little as 4 hours.

    Correlate siRNA Uptake with Gene Silencing
    Studies show that the silencing activity of Silencer™ Cy™3 Labeled GAPDH siRNA is not affected by the fluorescent label. Therefore, these siRNAs are ideal for examining both siRNA uptake and siRNA-induced knockdown, and for correlating gene knockdown with transfection efficiency; this is important to know because different transfection reagents interact with siRNAs in different ways (i.e., some reagents may sequester the siRNA, resulting in poor gene knockdown). The Silencer™ Cy™3 Labeled GAPDH siRNA has the fluorescein derivative Cy™3 (excitation max. = 547, emission max. = 563 nm) on the 5' end of the sense strand, making it highly fluorescent when excited by the appropriate wavelength of light. Silencer™ Cy™3 Labeled GAPDH siRNA is suitable for monitoring siRNA uptake by chemical transfection or electroporation. Cy™3 labeling is recommended over FAM™ labeling when using electroporation, because of its brighter fluorescence signal visible over a broader pH range. Electroporation produces a more diffuse siRNA distribution pattern, resulting in a less intense fluorescence signal than with chemical transfection.

    Accessory Products:
    Silencer™ Cy™3 Labeled GAPDH siRNA (human) (SKU #AM4623) is also available as Silencer™ FAM™ Labeled GAPDH siRNA for human (SKU #AM4622) and for human, mouse, and rat (SKU #AM4650). Silencer™ control siRNAs are unmodified 21-mers; they are designed to be used with Silencer™ siRNAs, BLOCK-iT™ siRNAs, and other unmodified 21mer siRNAs. If you are using other siRNA products, such as Invitrogen™ Silencer™ Select siRNAs or STEALTH RNAi™ siRNAs, we recommend that you use control siRNAs designed for use with those siRNAs for more reliable experimental results.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Content And Storage The siRNA is provided dried down in a single tube, along with Nuclease-free Water for resuspension, and should be stored at –20°C.
    RNAi Type siRNA
    Species Human, Mouse, Rat
    Label or Dye Cy3
    Product Line Silencer, Ambion
    Product Type siRNA
    Purity HPLC
    Quantity 5 nmol
    Shipping Condition Dry Ice
    Target GAPDH
    How critical is it to include positive and negative controls in siRNA experiments?

    A positive control siRNA such as the Silencer Select GAPDH siRNA will demonstrate efficient transfection and help to optimize transfection conditions. A negative control siRNA is necessary to serve as a baseline for gene expression.

    What are the positive and negative controls I should use with my Silencer or Silencer Select siRNA experiment?

    We offer several negative and positive controls for both Silencer and Silencer Select siRNA.

    For Silencer siRNA positive control:
    - Silencer Firefly Luciferase (GL2 + GL3) siRNA (Cat. No. AM4629, 5nmol)
    - Silencer GAPDH siRNA (human) (Cat. No AM4605, 5nmol; AM4633, 40 nmol)
    - Silencer GAPDH siRNA (human, mouse, rat) (Cat. No. AM4624, 5 nmol; AM4631, 40 nmol; AM4632, 5 x 40 nmol)
    - Silencer GFP (eGFP) siRNA (Cat. No. AM4626, 5 nmol)
    - Silencer KIF11 (Eg5) siRNA (human, mouse, rat) (Cat. No. AM4639, 5 nmol)

    If working with Fluorescently labeled Silencer siRNA:
    - Silencer Cy-3 labeled GAPDH siRNA (human, mouse, rat) (Cat. No. AM4649, 5 nmol)
    - Silencer FAM-labeled GAPDH siRNA (human, mouse, rat) (Cat. NO. AM4650, 5 nmol)

    If working with Silencer siRNA in vivo ready:
    - Silencer GAPDH Positive Control siRNA, in vivo ready (Cat. No. 4404025, 250 nmol)

    For Silencer siRNA negative controls:
    - Silencer Negative Control No. 1 siRNA (Cat. Nos. AM4611, 5 nmol; AM4635, 40 nmol; AM4636, 5 x 40 nmol)
    - Silencer Negative Control No. 2 siRNA (Cat. Nos. AM4613, 5 nmol; AM4637, 30 nmol)
    - Silencer Negative Control No. 3 siRNA (Cat. No. AM4615, 5 nmol)
    - Silencer Negative Control No. 4 siRNA (Cat. No. AM4641, 5 nmol)
    - Silencer Negative Control No. 5 siRNA (Cat. No. AM4642, 5 nmol)

    If working with Fluorescently labeled Silencer siRNA:
    - Silencer Cy 3-labeled Negative Control No. 1 siRNA (Cat. No AM4621, 5 nmol)
    - Silencer FAM-labeled Negative Control No. 1 siRNA (Cat. No. AM4620, 5 nmol)

    If working with Silencer siRNA in vivo ready:
    - Silencer in vivo ready (Cat. No. 4404021)

    For Silencer Select siRNA Positive control:
    - Silencer Select GAPDH Positive Control siRNA (Cat. No. 4390849, 5 nmol; 4390850, 40 nmol; 4404024, 250 nmol)
    - Silencer Select MALAT1 Positive Control siRNA for non-coding siRNA (Cat. No. 4455877, 5 nmol)

    For Silencer Select siRNA negative control:
    - Silencer Select Negative Control No. 1 siRNA (Cat. No. 4390843, 5 nmol; 4390844, 40 nmol)
    - Silencer Select Negative Control No. 2 siRNA (Cat. No. 4390846, 5 nmol; 4390847, 40 nmol)

    If working with Silencer Select siRNA in vivo ready:
    - Silencer Select Negative Control No. 1 siRNA, in vivo ready (Cat. No. 4404020, 250nmol)

    WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.