Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Silencer™ Select Human Cell Surface siRNA Library

Catalog No. A30144
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A30144 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A30144 Supplier Invitrogen™ Supplier No. A30144
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Cell surface receptors are specialized integral membrane proteins that take part in communication between the cell and its surroundings.

Cell surface receptors are specialized integral membrane proteins that take part in communication between the cell and its surroundings. Extracellular signaling molecules, typically hormones, neurotransmitters, cytokines, growth factors and cell recognition molecules, attach to the receptor, triggering changes in the function of the cell. The Silencer™ Select Human Cell Surface siRNA Library contains 2,382 Ambion™ Silencer Select siRNAs targeting 794 genes. This library of highly potent, chemically-modified siRNAs was designed using the most up-to-date genome databases, including the NCBI RefSeq database and cross-referenced to the Gene Ontology Consortium (GO) database and/or the HUGO Gene Nomenclature Committee (HGNC). The Silencer Select siRNAs are designed using Thermo Fisher Scientific's leading algorithms—along with our novel locked nucleic acid (LNA) chemical modifications, this helps to ensure the efficacy, specificity and potency required for RNAi screening against cell surface-based signaling.

  • Superior efficacy – Reduce off target effects by up to 90% due to latest design algorithms and LNA chemical modifications
  • Highest efficiency – Perform knockdown studies with 100-fold more potent siRNAs than currently available siRNAs
  • Dependable – Achieve faster discoveries via consistency of gene knockdown and reliability of phenotypic results
  • Convenient – Reduce data de-convolution and accelerate target validation with our arrayed format
  • Customizable – Choose the best option for your workflow demands, including additional siRNA scales and pooled formats

Why use the Silencer Select Human Cell Surface siRNA Library?
Together with careful bioinformatics selection of gene targets, Silencer Select siRNAs are optimized with latest design algorithms, proprietary chemical modifications and high quality synthesis to help ensure desired RNAi outcomes. The Silencer Select Human Cell Surface siRNA Library contains our pre-designed and when available, validated siRNAs generated with highest potency and specificity in mind so you can focus on the highest data quality backed by the best in industry guarantee.

Silencer Select siRNA libraries empower you with highly potent siRNAs to maximize discovery with even less
Ambion Silencer Select siRNAs are designed with the highest potency of available siRNAs, resulting in 100-fold more potent siRNAs and allowing gene knockdowns at the lowest concentrations (see figure below). The highly potent Silencer Select siRNAs allow you to use even lower concentrations of siRNAs, which helps to minimize off target effects, reduce false negative data and maximize siRNA library output for additional screens and data validation. The Silencer Select Human Cell Surface siRNA Library contains 0.25 nmol of each highly potent siRNA, sufficient for at least 500 transfections (at 5 nM siRNA, 100 μL transfection volume).

Flexibility to meet your workflow needs
Thermo Fisher Scientific offers a comprehensive list of pre-defined or custom siRNA libraries to human, mouse or rat genes with 1, 2, or 5 nmol of each siRNA or larger sizes. For a complete gene list, additional information or to discuss your research with one of our technical experts, contact your local sales representative or e-mail us at RNAilibraries@lifetech.com.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2382 total siRNAs targeting 794 genes plated onto 30 x 96-well plates

Store at or below -20°C in a non-frost-free freezer.
Format 96-well Plate
RNAi Type siRNA
Species Human
Sample Type RNA
For Use With (Application) RNAi
Product Line Silencer™Select
Product Type siRNA Library
Purity Standard
Quantity 1 Set
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.