Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streptococcus group A Rabbit anti-Streptococcus, Polyclonal, Abnova™

Catalog No. 89033659 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-033-659 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-033-659 Supplier Abnova Corporation Supplier No. PAB29814
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Rabbit polyclonal antibody raised against Streptococcus group A.

  • High specificity
  • High reactivity

Specifications

Antigen Streptococcus group A
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against Streptococcus group A.
Dilution ELISA The optimal working dilution should be determined by the end user.
Formulation In 0.02 M sodium phosphate, 0.15 M sodium chloride (0.09% sodium azide).
Host Species Rabbit
Immunogen Streptococcus pyogenes.
Purification Method Affinity purification
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody reacts with Streptococcus group A. No reactivity with tested strains of Streptococcus group B, C, F, G, S. aureus, H. influenzae, N. perflava.
Target Species Streptococcus species
Content And Storage Store at 4°C.
Form Liquid
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.