Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streptococcus group B Rabbit anti-Streptococcus, Polyclonal, Abnova™

Catalog No. 89033662 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-033-662 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-033-662 Supplier Abnova Corporation Supplier No. PAB29817
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Rabbit polyclonal antibody raised against Streptococcus group B.

  • High specificity
  • High reactivity

Specifications

Antigen Streptococcus group B
Applications ELISA, Immunofluorescence, Lateral Flow, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against Streptococcus group B.
Dilution ELISA Immunofluorescence Lateral-Flow Immunochromatographic Assay Western Blot The optimal working dilution should be determined by the end user.
Formulation In 0.01 M Phospate buffered saline, pH 7.2 (0.09% sodium azide).
Host Species Rabbit
Immunogen Carbohydrate of Streptococcus group B.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody reacts with Streptococcus group B. Does not cross-react with other Streptococcus groups.
Target Species Streptococcus species
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.