Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl Alcohol, ACS, 99.0%, MilliporeSigma™

Catalog No. 50948022 Shop All MilliporeSigma Products
Change view
Click to view available options
Packaging:
Plastic Bottle
Tinplate Metal Can
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging
50-948-022 Plastic Bottle
50-947-978 Tinplate Metal Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-948-022 Supplier MilliporeSigma Supplier No. BX18052
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 75-65-0
Molecular Weight (g/mol) 74.12

Specifications

Melting Point 24 to 25°C
Quantity 1 L
Flash Point 14°C
Packaging Plastic Bottle
Residue after Evaporation 0.003% max
Assay Percent Range 99.0% min.
Vapor Pressure 40.7 hPa (20°C)
Boiling Point 81°C to 83°C (1013 hPa)
Recommended Storage Store at room temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.