Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mirus Bio™ TransIT™-Lenti Transfection Reagent

Catalog No. MIR6604 Shop All Mirus Bio LLC Products
Change view
Click to view available options
Quantity:
0.3 mL
0.75 mL
1.5 mL
5 x 1.5 mL
10 x 1.5 mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MIR6604 0.75 mL
MIR6603 0.3 mL
MIR6600 1.5 mL
MIR6605 5 x 1.5 mL
MIR6606 10 x 1.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MIR6604 Supplier Mirus Bio™ Supplier No. MIR 6604
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TransIT™-Lenti Transfection Reagent is ideal for recombinant Lentivirus production.

TransIT™-Lenti Transfection Reagent is designed to enhance delivery of packaging and transfer vectors to adherent HEK 293T cell types and increase recombinant lentivirus production.

  • High performance: Provide up to eight-fold higher functional titers
  • Simple protocol: No media change required, single harvest
  • Animal origin free: Regulatory friendly

Recommended Storage

Store at -20°C. Product guaranteed 6 months from date of purchase.

Specifications

Content And Storage Store at -20°C
Description TransIT™-Lenti Transfection Reagent
Cell Type 293-derived Cells
Format Tube
Promoter None (Promoterless)
Transfection Technique Chemical Transfection
For Use With (Application) Transfection, Transduction, Gene expression, Lentivirus production
Product Type Transfection Reagent
Quantity 0.75 mL
Sterility Sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.